missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-picoline, 99%

Catalog No. p-57880
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104390100 10 g Glass bottle
AC104390500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104390100 Supplier Thermo Scientific Chemicals Supplier No. 104390100

CAS: 1603-41-4 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1603-41-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006328
InChI Key CMBSSVKZOPZBKW-UHFFFAOYSA-N
Synonym 2-amino-5-methylpyridine, 2-amino-5-picoline, 6-amino-3-picoline, 2-pyridinamine, 5-methyl, 5-methylpyridin-2-ylamine, 3-picoline, 6-amino, 5-methyl-2-pyridinamine, 5-methyl-2-pyridylamine, 5-methyl-2-aminopyridine, 2-amino-5-methyl pyridine
PubChem CID 15348
IUPAC Name 5-methylpyridin-2-amine
SMILES CC1=CN=C(C=C1)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 80.0°C to 81.0°C
Color White to Yellow
Boiling Point 227.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 22, V,9, 289
Solubility Information Solubility in water: freely soluble.
Formula Weight 108.14
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2-Amino-5-picoline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 216-503-5
RTECSNumber TJ5141000
TSCA TSCA

RUO – Research Use Only