missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-phenylpyridine 98.0+%, TCI America™
$88.56 - $550.61
Chemical Identifiers
| CAS | 33421-40-8 |
|---|---|
| Molecular Formula | C11H10N2 |
| Molecular Weight (g/mol) | 170.22 |
| MDL Number | MFCD01692452 |
| InChI Key | OAPVIBHQRYFYSE-UHFFFAOYSA-N |
| Synonym | 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine |
| PubChem CID | 105097 |
| IUPAC Name | 5-phenylpyridin-2-amine |
| SMILES | NC1=CC=C(C=N1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1667100MG
|
TCI America
A1667100MG |
100 mg |
Each for $88.56
|
|
|||||
|
A16671G
|
TCI America
A16671G |
1 g |
Each for $550.61
|
|
|||||
Chemical Identifiers
| 33421-40-8 | |
| 170.22 | |
| OAPVIBHQRYFYSE-UHFFFAOYSA-N | |
| 105097 | |
| NC1=CC=C(C=N1)C1=CC=CC=C1 |
| C11H10N2 | |
| MFCD01692452 | |
| 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine | |
| 5-phenylpyridin-2-amine |
Specifications
| 33421-40-8 | |
| Yellow | |
| MFCD01692452 | |
| 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine | |
| NC1=CC=C(C=N1)C1=CC=CC=C1 | |
| 170.22 | |
| 170.22 | |
| Crystalline Powder |
| 135°C | |
| C11H10N2 | |
| 100 mg | |
| OAPVIBHQRYFYSE-UHFFFAOYSA-N | |
| 5-phenylpyridin-2-amine | |
| 105097 | |
| ≥98.0% (GC,T) | |
| 2-Amino-5-phenylpyridine |
Safety and Handling
RTECSNumber : US2145000
TSCA : No