Learn More
2-Amino-5-phenylpyridine, 97%
CAS: 33421-40-8 | C11H10N2 | 170.22 g/mol
Supplier: Thermo Scientific Chemicals H2600203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Amino-5-phenylpyridine | |
| 133°C to 137°C | |
| MFCD01692452 | |
| 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine | |
| NC1=CC=C(C=N1)C1=CC=CC=C1 | |
| 170.22 | |
| 170.21 |
| 33421-40-8 | |
| C11H10N2 | |
| 1 g | |
| OAPVIBHQRYFYSE-UHFFFAOYSA-N | |
| 5-phenylpyridin-2-amine | |
| 105097 | |
| 97% |
Chemical Identifiers
| 33421-40-8 | |
| 170.22 | |
| OAPVIBHQRYFYSE-UHFFFAOYSA-N | |
| 105097 | |
| NC1=CC=C(C=N1)C1=CC=CC=C1 |
| C11H10N2 | |
| MFCD01692452 | |
| 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine | |
| 5-phenylpyridin-2-amine |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H351
RTECSNumber : US2145000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only