missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-phenyl-3-furonitrile, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals AW01254EB
| Quantity | 25g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 14742-32-6 | |
| 184.20 | |
| UWTDTJWGYWUILP-UHFFFAOYSA-N | |
| 693961 | |
| NC1=C(C=C(O1)C1=CC=CC=C1)C#N |
| C11H8N2O | |
| MFCD00463472 | |
| 2-amino-5-phenyl-3-furonitrile, 2-amino-3-cyano-5-phenylfuran, 2-amino-5-phenyl-3-furancarbonitrile, 3-furancarbonitrile, 2-amino-5-phenyl, maybridge1_000452, 2-amino-5-phenyl-furan-3-carbonitrile, 3-furancarbonitrile,2-amino-5-phenyl, 2-azanyl-5-phenyl-furan-3-carbonitrile | |
| 2-amino-5-phenylfuran-3-carbonitrile |
Specifications
| 2-Amino-5-phenyl-3-furonitrile | |
| 94.5 | |
| 95% | |
| C11H8N2O | |
| MFCD00463472 | |
| UWTDTJWGYWUILP-UHFFFAOYSA-N | |
| 2-amino-5-phenylfuran-3-carbonitrile | |
| 693961 | |
| 95% |
| 14742-32-6 | |
| 100.0 | |
| Amber Glass Bottle | |
| 25g | |
| 2-amino-5-phenyl-3-furonitrile, 2-amino-3-cyano-5-phenylfuran, 2-amino-5-phenyl-3-furancarbonitrile, 3-furancarbonitrile, 2-amino-5-phenyl, maybridge1_000452, 2-amino-5-phenyl-furan-3-carbonitrile, 3-furancarbonitrile,2-amino-5-phenyl, 2-azanyl-5-phenyl-furan-3-carbonitrile | |
| NC1=C(C=C(O1)C1=CC=CC=C1)C#N | |
| 184.20 | |
| 184.2 |
Safety and Handling
GHS Signal Word: Warning