missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrothiazole 98.0+%, TCI America™

$163.64 - $163.64

Chemical Identifiers

CAS 121-66-4
Molecular Formula C3H3N3O2S
Molecular Weight (g/mol) 145.14
MDL Number MFCD00005326
InChI Key MIHADVKEHAFNPG-UHFFFAOYSA-N
Synonym 2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
PubChem CID 8486
ChEBI CHEBI:82386
IUPAC Name 5-nitro-1,3-thiazol-2-amine
SMILES NC1=NC=C(S1)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A038125G
View Documents
TCI America
A038125G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

121-66-4
145.14
MIHADVKEHAFNPG-UHFFFAOYSA-N
8486
5-nitro-1,3-thiazol-2-amine
C3H3N3O2S
MFCD00005326
2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
CHEBI:82386
NC1=NC=C(S1)[N+]([O-])=O

Specifications

121-66-4
C3H3N3O2S
25 g
MIHADVKEHAFNPG-UHFFFAOYSA-N
5-nitro-1,3-thiazol-2-amine
8486
145.14
Crystalline Powder
202°C
MFCD00005326
2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
NC1=NC=C(S1)[N+]([O-])=O
145.14
CHEBI:82386
≥98.0% (HPLC,T)
2-Amino-5-nitrothiazole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0206

RTECSNumber : XJ2800000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'