missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrothiazole, 97%

Catalog No. p-7025699
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2133514 25 g
AAB2133522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2133514 Supplier Thermo Scientific Chemicals Supplier No. B2133514
Only null left

CAS: 121-66-4 | C3H3N3O2S | 145.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-66-4
Molecular Formula C3H3N3O2S
Molecular Weight (g/mol) 145.14
MDL Number MFCD00005326
InChI Key MIHADVKEHAFNPG-UHFFFAOYSA-N
Synonym 2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
PubChem CID 8486
ChEBI CHEBI:82386
IUPAC Name 5-nitro-1,3-thiazol-2-amine
SMILES NC1=NC=C(S1)[N+]([O-])=O

Specifications

Melting Point ∼202°C (decomposition)
Quantity 25 g
Beilstein 126797
Sensitivity Light sensitive
Merck Index 14,457
Formula Weight 145.14
Percent Purity 97%
Chemical Name or Material 2-Amino-5-nitrothiazole
Hazard Category H302+H312+H332-H315-H319-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-490-9
RTECSNumber XJ2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only