Learn More
2-Amino-5-nitrophenol, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 400900500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2-Amino-5-nitrophenol | |
| 121-88-0 | |
| 100.0 | |
| 95% | |
| C6H6N2O3 | |
| 50g | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and diethylether | |
| C1=CC(=C(C=C1[N+](=O)[O-])O)N | |
| 154.13 | |
| CHEBI:82384 | |
| 95% |
| 95% | |
| 94.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00007692 | |
| 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene | |
| DOPJTDJKZNWLRB-UHFFFAOYSA-N | |
| 2-amino-5-nitrophenol | |
| 4984721 | |
| 154.13 |
Chemical Identifiers
| 121-88-0 | |
| 154.13 | |
| DOPJTDJKZNWLRB-UHFFFAOYSA-N | |
| 4984721 | |
| 2-amino-5-nitrophenol |
| C6H6N2O3 | |
| MFCD00007692 | |
| 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene | |
| CHEBI:82384 | |
| C1=CC(=C(C=C1[N+](=O)[O-])O)N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Warning
EINECSNumber : 204-503-8
RTECSNumber : SJ6302500
TSCA : TSCA
RUO â Research Use Only