missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 400900010

 View more versions of this product

Catalog No. AC400900010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Amino-5-nitrophenol
121-88-0
100.0
95%
C6H6N2O3
1kg
Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and diethylether
C1=CC(=C(C=C1[N+](=O)[O-])O)N
154.13
CHEBI:82384
95%
95%
94.0
Authentic
Glass bottle
MFCD00007692
2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
DOPJTDJKZNWLRB-UHFFFAOYSA-N
2-amino-5-nitrophenol
4984721
154.13

Chemical Identifiers

121-88-0
154.13
DOPJTDJKZNWLRB-UHFFFAOYSA-N
4984721
2-amino-5-nitrophenol
C6H6N2O3
MFCD00007692
2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
CHEBI:82384
C1=CC(=C(C=C1[N+](=O)[O-])O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice

GHS Signal Word: Warning

EINECSNumber : 204-503-8

RTECSNumber : SJ6302500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only