missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylthiazole, 98%, Thermo Scientific™

Chemical Identifiers

CAS 7305-71-7
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00078317
InChI Key GUABFMPMKJGSBQ-UHFFFAOYSA-N
Synonym 2-amino-5-methylthiazole, 5-methylthiazol-2-amine, 2-thiazolamine, 5-methyl, 2-amino-5-methyl-thiazole, thiazole, 2-amino-5-methyl, 5-methyl-2-thiazolamine, 5-methyl-2-aminothiazole, unii-7rab78af2p, 2-amino-5-methyl thiazole, meloxicam related compound b
PubChem CID 351770
IUPAC Name 5-methyl-1,3-thiazol-2-amine
SMILES CC1=CN=C(S1)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC392790050
SDS View Documents
Thermo Scientific Chemicals
392790050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC392790250
SDS View Documents
Thermo Scientific Chemicals
392790250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC392791000
SDS View Documents
Thermo Scientific Chemicals
392791000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7305-71-7
114.17
GUABFMPMKJGSBQ-UHFFFAOYSA-N
351770
CC1=CN=C(S1)N
C4H6N2S
MFCD00078317
2-amino-5-methylthiazole, 5-methylthiazol-2-amine, 2-thiazolamine, 5-methyl, 2-amino-5-methyl-thiazole, thiazole, 2-amino-5-methyl, 5-methyl-2-thiazolamine, 5-methyl-2-aminothiazole, unii-7rab78af2p, 2-amino-5-methyl thiazole, meloxicam related compound b
5-methyl-1,3-thiazol-2-amine

Specifications

7305-71-7
100.0
97.5% min. (GC)
C4H6N2S
5g
GUABFMPMKJGSBQ-UHFFFAOYSA-N
5-methyl-1,3-thiazol-2-amine
351770
98%
97.5
Authentic
Glass bottle
MFCD00078317
2-amino-5-methylthiazole, 5-methylthiazol-2-amine, 2-thiazolamine, 5-methyl, 2-amino-5-methyl-thiazole, thiazole, 2-amino-5-methyl, 5-methyl-2-thiazolamine, 5-methyl-2-aminothiazole, unii-7rab78af2p, 2-amino-5-methyl thiazole, meloxicam related compound b
CC1=CN=C(S1)N
114.17
114.17
2-Amino-5-methylthiazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only