missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylthiazole, 98%

Catalog No. AC392791000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392791000 100 g Glass bottle
AC392790050 5 g Glass bottle
AC392790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC392791000 Supplier Thermo Scientific Chemicals Supplier No. 392791000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7305-71-7
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00078317
InChI Key GUABFMPMKJGSBQ-UHFFFAOYSA-N
Synonym 2-amino-5-methylthiazole, 5-methylthiazol-2-amine, 2-thiazolamine, 5-methyl, 2-amino-5-methyl-thiazole, thiazole, 2-amino-5-methyl, 5-methyl-2-thiazolamine, 5-methyl-2-aminothiazole, unii-7rab78af2p, 2-amino-5-methyl thiazole, meloxicam related compound b
PubChem CID 351770
IUPAC Name 5-methyl-1,3-thiazol-2-amine
SMILES CC1=CN=C(S1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 114.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-5-methylthiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only