missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylpyridine 98.0+%, TCI America™

Catalog No. p-7117034
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A073225G 25 g
A0732500G 500 g
2 options

Catalog No. A073225G

Supplier: TCI America A073225G

Only null left

Chemical Identifiers

CAS 1603-41-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006328
InChI Key CMBSSVKZOPZBKW-UHFFFAOYSA-N
Synonym 2-amino-5-methylpyridine, 2-amino-5-picoline, 6-amino-3-picoline, 2-pyridinamine, 5-methyl, 5-methylpyridin-2-ylamine, 3-picoline, 6-amino, 5-methyl-2-pyridinamine, 5-methyl-2-pyridylamine, 5-methyl-2-aminopyridine, 2-amino-5-methyl pyridine
PubChem CID 15348
IUPAC Name 5-methylpyridin-2-amine
SMILES CC1=CN=C(C=C1)N

Specifications

Melting Point 78°C
Boiling Point 227°C
Quantity 25 g
UN Number 2811
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methylpyridine
RTECSNumber TJ5141000
TSCA Yes