missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylpyridine, 99%

Catalog No. p-7021125
Change view
Click to view available options
Quantity:
50 g
250 g
Catalog No. Quantity
AAA1333018 50 g
AAA1333030 250 g
2 options

Catalog No. AAA1333018

Supplier: Thermo Scientific Chemicals A1333018

Only null left

CAS: 1603-41-4 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1603-41-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006328
InChI Key CMBSSVKZOPZBKW-UHFFFAOYSA-N
Synonym 2-amino-5-methylpyridine, 2-amino-5-picoline, 6-amino-3-picoline, 2-pyridinamine, 5-methyl, 5-methylpyridin-2-ylamine, 3-picoline, 6-amino, 5-methyl-2-pyridinamine, 5-methyl-2-pyridylamine, 5-methyl-2-aminopyridine, 2-amino-5-methyl pyridine
PubChem CID 15348
IUPAC Name 5-methylpyridin-2-amine
SMILES CC1=CN=C(C=C1)N

Specifications

Melting Point 74°C to 78°C
Boiling Point 226°C to 228°C
Flash Point 118°C (244°F)
Odor Unpleasant
Quantity 50 g
UN Number UN2811
Beilstein 107050
Sensitivity Hygroscopic
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2-Amino-5-methylpyridine
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 216-503-5
RTECSNumber TJ5141000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only