missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 160200010

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC160200010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2941-78-8
151.17
NBUUUJWWOARGNW-UHFFFAOYSA-N
76255
CC1=CC=C(N)C(=C1)C(O)=O
C8H9NO2
MFCD00007909
5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
2-amino-5-methylbenzoic acid

Specifications

2-Amino-5-methylbenzoic acid
96.0
Authentic
Glass bottle
H2NC6H3(CH3)CO2H
MFCD00007909
5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
CC1=CC=C(N)C(=C1)C(O)=O
151.17
151.16
2941-78-8
100.0
96% min. (HPLC)
C8H9NO2
1g
14, 481
NBUUUJWWOARGNW-UHFFFAOYSA-N
2-amino-5-methylbenzoic acid
76255
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-932-3

SDS
Documents

Documents

Promotions

Promotions