missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methyl-1,3,4-thiadiazole 98.0+%, TCI America™

Catalog No. A08745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08745G 5 g
A087425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08745G Supplier TCI America Supplier No. A08745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-33-8
Molecular Formula C3H5N3S
Molecular Weight (g/mol) 115.15
MDL Number MFCD00003110
InChI Key HMPUHXCGUHDVBI-UHFFFAOYSA-N
Synonym 2-amino-5-methyl-1,3,4-thiadiazole, 2-amino-5-methylthiadiazole, matd, usaf cy-3, 1,3,4-thiadiazol-2-amine, 5-methyl, 5-methyl-1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazole, 2-amino-5-methyl, 5-methyl-1,3,4-thiadiazole-2-amine, 5-methyl-1,3,4-thiadiazol-2-ylamine, 1,4-thiadiazol-2-amine, 5-methyl
PubChem CID 66949
IUPAC Name 5-methyl-1,3,4-thiadiazol-2-amine
SMILES CC1=NN=C(N)S1

Specifications

Color White
Quantity 5 g
Formula Weight 115.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methyl-1,3,4-thiadiazole
RTECSNumber XI3500000
TSCA No