missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methoxybenzoic acid, 97%

Catalog No. AC443340050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC443340050 5 g Glass bottle
AC443340010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443340050 Supplier Thermo Scientific Chemicals Supplier No. 443340050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6705-03-9 | C11H13N3O | 203.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6705-03-9

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 151°C to 151.7°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C11H13N3O
Quantity 5 g
Synonym 2-amino-3-1h-indol-3-yl propanamide, h-dl-trp-nh2, alpha-amino-1h-indole-3-propanamide, tryptophan amide, dl-tryptophanamide, d,l-tryptophanamide, l-tryptophanamide, 9, h-dl-trp-nh hcl, dl-trytophan amide hydrochloride, 1h-indole-3-propanamide,a-amino
InChI Key JLSKPBDKNIXMBS-UHFFFAOYNA-N
SMILES NC(CC1=CNC2=CC=CC=C12)C(N)=O
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanamide
Molecular Weight (g/mol) 203.25
PubChem CID 89366
Formula Weight 167.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methoxybenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only