missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methoxybenzoic acid, 97%

Catalog No. AC443340010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC443340010 1 g Glass bottle
AC443340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443340010 Supplier Thermo Scientific Chemicals Supplier No. 443340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6705-03-9 | C11H13N3O | 203.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6705-03-9

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 151°C to 151.7°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H9NO3
Quantity 1 g
Synonym 5-methoxyanthranilic acid, 2-amino-5-methoxybenzoicacid, 2-amino-5-methoxy-benzoic acid, benzoic acid, 2-amino-5-methoxy, pubchem2544, acmc-1b7jp, 5-methoxy anthranilic acid, 5-methoxyanththranillic acid, ksc495k4p, anthranilic acid, 5-methoxy
InChI Key UMKSAURFQFUULT-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)N)C(=O)O
IUPAC Name 2-amino-5-methoxybenzoic acid
Molecular Weight (g/mol) 167.16
PubChem CID 277930
Formula Weight 167.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methoxybenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only