missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodopyridine 98.0+%, TCI America™

Catalog No. p-7117600
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18421G 1 g
A18425G 5 g
A184225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A18421G Supplier TCI America Supplier No. A18421G

Chemical Identifiers

CAS 20511-12-0
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.013
InChI Key IVILGUFRMDBUEQ-UHFFFAOYSA-N
Synonym 2-amino-5-iodopyridine, 2-pyridinamine, 5-iodo, 5-iodo-pyridin-2-ylamine, 5-iodo-2-pyridinamine, 5-iodo-2-aminopyridine, 2-amino-5-iodo pyridine, pyridine, 2-amino-5-iodo, 5-iod-2-pyridinamin, 5-iodo-2-pyridylamine, 6-amino-3-iodopyridine
PubChem CID 88579
IUPAC Name 5-iodopyridin-2-amine
SMILES C1=CC(=NC=C1I)N

Specifications

Melting Point 127°C
Quantity 1 g
Formula Weight 220.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-iodopyridine
TSCA Yes
Recommended Storage Refrigerator