missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodopyridine, 97%

CAS: 20511-12-0 | C5H5IN2 | 220.013 g/mol

Supplier:  Thermo Scientific Chemicals B2517506

 View more versions of this product

Catalog No. AAB2517506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-5-iodopyridine
126°C to 130°C
MFCD00160312
108738
2-amino-5-iodopyridine, 2-pyridinamine, 5-iodo, 5-iodo-pyridin-2-ylamine, 5-iodo-2-pyridinamine, 5-iodo-2-aminopyridine, 2-amino-5-iodo pyridine, pyridine, 2-amino-5-iodo, 5-iod-2-pyridinamin, 5-iodo-2-pyridylamine, 6-amino-3-iodopyridine
C1=CC(=NC=C1I)N
220.013
220.01
20511-12-0
C5H5IN2
5 g
Light sensitive
IVILGUFRMDBUEQ-UHFFFAOYSA-N
5-iodopyridin-2-amine
88579
97%

Chemical Identifiers

20511-12-0
220.013
IVILGUFRMDBUEQ-UHFFFAOYSA-N
88579
C1=CC(=NC=C1I)N
C5H5IN2
MFCD00160312
2-amino-5-iodopyridine, 2-pyridinamine, 5-iodo, 5-iodo-pyridin-2-ylamine, 5-iodo-2-pyridinamine, 5-iodo-2-aminopyridine, 2-amino-5-iodo pyridine, pyridine, 2-amino-5-iodo, 5-iod-2-pyridinamin, 5-iodo-2-pyridylamine, 6-amino-3-iodopyridine
5-iodopyridin-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only