missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodo-4-methylpyridine 98.0+%, TCI America™

Catalog No. p-7118124
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28801G 1 g
A28805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28801G Supplier TCI America Supplier No. A28801G

Chemical Identifiers

CAS 356561-08-5
Molecular Formula C6H7IN2
Molecular Weight (g/mol) 234.04
MDL Number MFCD06637727
InChI Key HYRZCIXFHXERJT-UHFFFAOYSA-N
Synonym 2-amino-5-iodo-4-methylpyridine, 5-iodo-4-methyl-pyridin-2-ylamine, 5-iodo-4-methylpyridin-2-ylamine, 2-pyridinamine, 5-iodo-4-methyl, 5-iodo-4-methyl-2-pyridinamine, 5-iodo-4-methyl-2-pyridylamine, 2-amino-5-iodo-4-picoline, pubchem9289, acmc-1ad4a, 2-amino-4-methyl-5-iodopyridine
PubChem CID 17750195
IUPAC Name 5-iodo-4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1I)N

Specifications

Melting Point 121°C
Quantity 1 g
Formula Weight 234.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-iodo-4-methylpyridine