missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. AAH62575MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62575MD 250 mg
AAH6257503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62575MD Supplier Thermo Scientific Chemicals Supplier No. H62575MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 863578-24-9 | C12H17BFNO2 | 237.081 g/mol

2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 863578-24-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.081
MDL Number MFCD11617916
InChI Key RLUKWTHMONYTKG-UHFFFAOYSA-N
Synonym 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-fluorophenyl boronic acid pinacol ester, benzenamine,4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-amino-5-fluorobenzeneboronic acid pinacol ester, 4-fluoro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-fluoranyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 23152508
IUPAC Name 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)N

Specifications

Melting Point 49°C to 50°C
Quantity 250 mg
Formula Weight 237.08
Percent Purity 96%
Chemical Name or Material 2-Amino-5-fluorobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only