missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluoro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog No. p-7053688
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6129306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6129306 Supplier Thermo Scientific Chemicals Supplier No. H6129306
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 212268-12-7
Molecular Formula C5H4FN3O2
Molecular Weight (g/mol) 157.10
MDL Number MFCD04972413
InChI Key LDYYBZNEWDTDEE-UHFFFAOYSA-N
Synonym 2-amino-3-nitro-5-fluoropyridine, 2-amino-5-fluoro-3-nitropyridine, 5-fluoro-3-nitro-2-pyridinamine, pubchem1271, acmc-1cegr, ksc496c2h, 5-fluoro-3-nitro-2-aminopyridine, 5-fluoro-3-nitropyridine-2-amine, 5-fluoro-3-nitro-pyridin-2-amine, 5-fluoro-3-nitro-2-pyridylamine
PubChem CID 21717339
IUPAC Name 5-fluoro-3-nitropyridin-2-amine
SMILES NC1=NC=C(F)C=C1[N+]([O-])=O

Specifications

Color Orange
Quantity 5 g
Formula Weight 157.1
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 2-Amino-5-fluoro-3-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only