missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyrimidine 98.0+%, TCI America™

Catalog No. A19185G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19185G Supplier TCI America Supplier No. A19185G
Only null left

Chemical Identifiers

CAS 5428-89-7
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00014604
InChI Key OQZMDDKDHRIGDY-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyrimidine, 2-pyrimidinamine, 5-chloro, 5-chloro-2-pyrimidinamine, pyrimidine, 2-amino-5-chloro, 5-chloropyrimidine-2-ylamine, 5-chlorpyrimidin-2-amin, chloropyrimidinamine, zlchem 634, pubchem9619, acmc-1aya8
PubChem CID 79479
IUPAC Name 5-chloropyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)Cl

Specifications

Melting Point 238°C
Color White-Yellow
Quantity 5 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloropyrimidine
RTECSNumber UV6329500
TSCA Yes