missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorophenylboronic acid pinacol ester, 97%

Catalog No. AC444470010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444470010 1 g
AC444470050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444470010 Supplier Thermo Scientific Chemicals Supplier No. 444470010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1073371-77-3 | C12H17BClNO2 | 253.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073371-77-3
Molecular Formula C12H17BClNO2
Molecular Weight (g/mol) 253.53
MDL Number MFCD06795672
InChI Key GVJZHGCVSYBPIM-UHFFFAOYSA-N
Synonym 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid, pinacol ester, 2-amino-5-chlorophenylboronic acid pinacol ester, 4-chloro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid,pinacol ester, 2-amino-5-chlorobenzeneboronic acid, pinacol ester, 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine
PubChem CID 17750205
IUPAC Name 4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1

Specifications

Melting Point 91°C to 93°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 253.54
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorophenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

RUO – Research Use Only