missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzoxazole, 97%

Catalog No. AC103380250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103380250 25 g Glass bottle
AC103380010 1 g Glass bottle
AC103380050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103380250 Supplier Thermo Scientific Chemicals Supplier No. 103380250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61-80-3 | C7H5ClN2O | 168.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61-80-3
Molecular Formula C7H5ClN2O
Molecular Weight (g/mol) 168.58
MDL Number MFCD00005770
InChI Key YGCODSQDUUUKIV-UHFFFAOYSA-N
Synonym zoxazolamine, 2-amino-5-chlorobenzoxazole, 5-chlorobenzo d oxazol-2-amine, flexin, 2-benzoxazolamine, 5-chloro, contrazole, deflexol, flexilon, zoxamin, zoxine
PubChem CID 6103
ChEBI CHEBI:35053
IUPAC Name 5-chloro-1,3-benzoxazol-2-amine
SMILES NC1=NC2=CC(Cl)=CC=C2O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180.0°C to 185.0°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 10399
Formula Weight 168.58
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-chlorobenzoxazole, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-519-4
RTECSNumber DM4550000

RUO – Research Use Only