missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7117522
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15725G 5 g
A157225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15725G Supplier TCI America Supplier No. A15725G
Only null left

Chemical Identifiers

CAS 5922-60-1
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00017106
InChI Key QYRDWARBHMCOAG-UHFFFAOYSA-N
Synonym 5-chloroanthranilonitrile, benzonitrile, 2-amino-5-chloro, anthranilonitrile, 5-chloro, 2-amino-5-chloro-benzonitrile, 4-chloro-2-cyanoaniline, unii-gy0x0082qb, 2-amino-5-chloro benzonitrile, amino 2-5-chloro-benzonitrile, 2-amino-5-chlorobenzenecarbonitrile, acbn
PubChem CID 80019
IUPAC Name 2-amino-5-chlorobenzonitrile
SMILES NC1=CC=C(Cl)C=C1C#N

Specifications

Melting Point 96°C
Color White-Yellow
Boiling Point 159°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 152.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Amino-5-chlorobenzonitrile