missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzonitrile, 97%

Catalog No. p-7026608
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2269003 1 g
AAB2269006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2269003 Supplier Thermo Scientific Chemicals Supplier No. B2269003
Only null left

CAS: 5922-60-1 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5922-60-1
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00017106
InChI Key QYRDWARBHMCOAG-UHFFFAOYSA-N
Synonym 5-chloroanthranilonitrile, benzonitrile, 2-amino-5-chloro, anthranilonitrile, 5-chloro, 2-amino-5-chloro-benzonitrile, 4-chloro-2-cyanoaniline, unii-gy0x0082qb, 2-amino-5-chloro benzonitrile, amino 2-5-chloro-benzonitrile, 2-amino-5-chlorobenzenecarbonitrile, acbn
PubChem CID 80019
IUPAC Name 2-amino-5-chlorobenzonitrile
SMILES NC1=CC=C(Cl)C=C1C#N

Specifications

Melting Point 94°C to 97°C
Boiling Point 132°C to 135°C (0.5 mmHg)
Quantity 1 g
UN Number UN3439
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 227-651-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only