missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzenesulfonic acid, 97%

Catalog No. AAB2573814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2573814 25 g
AAB2573806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2573814 Supplier Thermo Scientific Chemicals Supplier No. B2573814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 133-74-4 | C6H6ClNO3S | 207.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 133-74-4
Molecular Formula C6H6ClNO3S
Molecular Weight (g/mol) 207.63
MDL Number MFCD00035776
InChI Key ZCGVPUAAMCMLTM-UHFFFAOYSA-N
Synonym 2-amino-5-chlorobenzenesulphonic acid, 5-chloroorthanilic acid, 4-chloroaniline-2-sulfonic acid, 4-chloro-2-sulphoaniline, p-chloranilin-o-sulfosaure, p-chloroaniline-2-sulphonic acid, 2-amino-5-chloro-benzenesulfonic acid, benzenesulfonic acid,2-amino-5-chloro, 2-amino-5-chlorobenzene-1-sulfonic acid, 2-azanyl-5-chloranyl-benzenesulfonic acid
PubChem CID 8628
IUPAC Name 2-amino-5-chlorobenzenesulfonic acid
SMILES NC1=CC=C(Cl)C=C1S(O)(=O)=O

Specifications

Melting Point >300°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 207.64
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chlorobenzenesulfonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-119-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only