missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-3-iodopyridine 98.0+%, TCI America™

Catalog No. A30695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30695G 5 g
A30691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30695G Supplier TCI America Supplier No. A30695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 211308-81-5
Molecular Formula C5H4ClIN2
Molecular Weight (g/mol) 254.455
MDL Number MFCD07644578
InChI Key DIONPYCYVWCDIG-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-3-iodopyridine, 5-chloro-3-iodo-pyridin-2-ylamine, 5-chloro-3-iodo-2-pyridinamine, 2-pyridinamine, 5-chloro-3-iodo, pubchem16614, acmc-1cc0x, ksc496m8l, 2-amino-5-chloro-3-iodopyridin, 5-chloro-3-iodo-2-pyridylamine, 5-chloro-3-iodo-2-aminopyridine
PubChem CID 11076010
IUPAC Name 5-chloro-3-iodopyridin-2-amine
SMILES C1=C(C=NC(=C1I)N)Cl

Specifications

Melting Point 98°C
Quantity 5 g
Formula Weight 254.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloro-3-iodopyridine