missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-3-iodopyridine, 95%, Thermo Scientific™

$149.16 - $149.16

Chemical Identifiers

CAS 211308-81-5
Molecular Formula C5H4ClIN2
Molecular Weight (g/mol) 254.455
MDL Number MFCD07644578
InChI Key DIONPYCYVWCDIG-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-3-iodopyridine, 5-chloro-3-iodo-pyridin-2-ylamine, 5-chloro-3-iodo-2-pyridinamine, 2-pyridinamine, 5-chloro-3-iodo, pubchem16614, acmc-1cc0x, ksc496m8l, 2-amino-5-chloro-3-iodopyridin, 5-chloro-3-iodo-2-pyridylamine, 5-chloro-3-iodo-2-aminopyridine
PubChem CID 11076010
IUPAC Name 5-chloro-3-iodopyridin-2-amine
SMILES C1=C(C=NC(=C1I)N)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2705303
View Documents
Thermo Scientific Chemicals
H2705303
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

211308-81-5
254.455
DIONPYCYVWCDIG-UHFFFAOYSA-N
11076010
C1=C(C=NC(=C1I)N)Cl
C5H4ClIN2
MFCD07644578
2-amino-5-chloro-3-iodopyridine, 5-chloro-3-iodo-pyridin-2-ylamine, 5-chloro-3-iodo-2-pyridinamine, 2-pyridinamine, 5-chloro-3-iodo, pubchem16614, acmc-1cc0x, ksc496m8l, 2-amino-5-chloro-3-iodopyridin, 5-chloro-3-iodo-2-pyridylamine, 5-chloro-3-iodo-2-aminopyridine
5-chloro-3-iodopyridin-2-amine

Specifications

211308-81-5
C5H4ClIN2
1 g
2-amino-5-chloro-3-iodopyridine, 5-chloro-3-iodo-pyridin-2-ylamine, 5-chloro-3-iodo-2-pyridinamine, 2-pyridinamine, 5-chloro-3-iodo, pubchem16614, acmc-1cc0x, ksc496m8l, 2-amino-5-chloro-3-iodopyridin, 5-chloro-3-iodo-2-pyridylamine, 5-chloro-3-iodo-2-aminopyridine
C1=C(C=NC(=C1I)N)Cl
254.455
254.46
2-Amino-5-chloro-3-iodopyridine
95°C to 98°C
MFCD07644578
Light sensitive
DIONPYCYVWCDIG-UHFFFAOYSA-N
5-chloro-3-iodopyridin-2-amine
11076010
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents