missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-3-iodopyridine, 95%, Thermo Scientific™

Catalog No. AAH2705303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2705303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2705303 Supplier Thermo Scientific Chemicals Supplier No. H2705303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 211308-81-5
Molecular Formula C5H4ClIN2
Molecular Weight (g/mol) 254.455
MDL Number MFCD07644578
InChI Key DIONPYCYVWCDIG-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-3-iodopyridine, 5-chloro-3-iodo-pyridin-2-ylamine, 5-chloro-3-iodo-2-pyridinamine, 2-pyridinamine, 5-chloro-3-iodo, pubchem16614, acmc-1cc0x, ksc496m8l, 2-amino-5-chloro-3-iodopyridin, 5-chloro-3-iodo-2-pyridylamine, 5-chloro-3-iodo-2-aminopyridine
PubChem CID 11076010
IUPAC Name 5-chloro-3-iodopyridin-2-amine
SMILES C1=C(C=NC(=C1I)N)Cl

Specifications

Melting Point 95°C to 98°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 254.46
Percent Purity 95%
Chemical Name or Material 2-Amino-5-chloro-3-iodopyridine
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures