missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-bromothiazole Hydrobromide 95.0+%, TCI America™
Supplier: TCI America A157325G
Specifications
| 2-Amino-5-bromothiazole Hydrobromide | |
| 163°C | |
| MFCD00012712 | |
| 2-amino-5-bromothiazole hydrobromide, 2-amino-5-bromothiazole monohydrobromide, 5-bromothiazol-2-amine hydrobromide, 5-bromo-1,3-thiazol-2-amine hydrobromide, 5-bromo-thiazol-2-ylamine hydrobromide, 2-amino-5-bromothiazole hbr, 2-amino-5-bromo-1,3-thiazole hydrobromide, 2-thiazolamine, 5-bromo-, monohydrobromide, 5-bromo-1,3-thiazole-2-ylamine, bromide, acmc-209mr7 | |
| C1=C(SC(=N1)N)Br.Br | |
| 259.947 | |
| 259.95 | |
| Crystalline Powder |
| 61296-22-8 | |
| C3H4Br2N2S | |
| 25 g | |
| NUSVDASTCPBUIP-UHFFFAOYSA-N | |
| 5-bromo-1,3-thiazol-2-amine;hydrobromide | |
| 2723848 | |
| ≥95.0% (T) |
Chemical Identifiers
| 61296-22-8 | |
| 259.947 | |
| NUSVDASTCPBUIP-UHFFFAOYSA-N | |
| 2723848 | |
| C1=C(SC(=N1)N)Br.Br |
| C3H4Br2N2S | |
| MFCD00012712 | |
| 2-amino-5-bromothiazole hydrobromide, 2-amino-5-bromothiazole monohydrobromide, 5-bromothiazol-2-amine hydrobromide, 5-bromo-1,3-thiazol-2-amine hydrobromide, 5-bromo-thiazol-2-ylamine hydrobromide, 2-amino-5-bromothiazole hbr, 2-amino-5-bromo-1,3-thiazole hydrobromide, 2-thiazolamine, 5-bromo-, monohydrobromide, 5-bromo-1,3-thiazole-2-ylamine, bromide, acmc-209mr7 | |
| 5-bromo-1,3-thiazol-2-amine;hydrobromide |
Safety and Handling
RTECSNumber : XJ1262200
TSCA : No