missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromothiazole Hydrobromide 95.0+%, TCI America™

Catalog No. A15735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15735G 5 g
A157325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15735G Supplier TCI America Supplier No. A15735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61296-22-8
Molecular Formula C3H4Br2N2S
Molecular Weight (g/mol) 259.947
MDL Number MFCD00012712
InChI Key NUSVDASTCPBUIP-UHFFFAOYSA-N
Synonym 2-amino-5-bromothiazole hydrobromide, 2-amino-5-bromothiazole monohydrobromide, 5-bromothiazol-2-amine hydrobromide, 5-bromo-1,3-thiazol-2-amine hydrobromide, 5-bromo-thiazol-2-ylamine hydrobromide, 2-amino-5-bromothiazole hbr, 2-amino-5-bromo-1,3-thiazole hydrobromide, 2-thiazolamine, 5-bromo-, monohydrobromide, 5-bromo-1,3-thiazole-2-ylamine, bromide, acmc-209mr7
PubChem CID 2723848
IUPAC Name 5-bromo-1,3-thiazol-2-amine;hydrobromide
SMILES C1=C(SC(=N1)N)Br.Br

Specifications

Melting Point 163°C
Quantity 5 g
Formula Weight 259.95
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromothiazole Hydrobromide
RTECSNumber XJ1262200
TSCA No