missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-bromopyrimidine 98.0+%, TCI America™
$76.43 - $320.62
Chemical Identifiers
| CAS | 7752-82-1 |
|---|---|
| Molecular Formula | C4H4BrN3 |
| Molecular Weight (g/mol) | 174.001 |
| InChI Key | UHRHPPKWXSNZLR-UHFFFAOYSA-N |
| Synonym | 2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine |
| PubChem CID | 231310 |
| IUPAC Name | 5-bromopyrimidin-2-amine |
| SMILES | C1=C(C=NC(=N1)N)Br |
Chemical Identifiers
| 7752-82-1 | |
| 174.001 | |
| 2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine | |
| 5-bromopyrimidin-2-amine |
| C4H4BrN3 | |
| UHRHPPKWXSNZLR-UHFFFAOYSA-N | |
| 231310 | |
| C1=C(C=NC(=N1)N)Br |
Specifications
| 7752-82-1 | |
| Yellow | |
| 5 g | |
| 2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine | |
| C1=C(C=NC(=N1)N)Br | |
| 174.001 | |
| 174.00 | |
| Crystalline Powder |
| 244°C | |
| C4H4BrN3 | |
| 3077 | |
| UHRHPPKWXSNZLR-UHFFFAOYSA-N | |
| 5-bromopyrimidin-2-amine | |
| 231310 | |
| ≥98.0% (GC,T) | |
| 2-Amino-5-bromopyrimidine |
Safety and Handling
TSCA : No