missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyridine, 97%

CAS: 1072-97-5 | C5H5BrN2 | 173.01 g/mol

$59.90 - $59.90

Chemical Identifiers

CAS 1072-97-5
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD00006323
InChI Key WGOLHUGPTDEKCF-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyridine, 2-pyridinamine, 5-bromo, 5-bromo-2-pyridylamine, 5-bromo-2-aminopyridine, 5-bromo-2-pyridinamine, 5-bromopyridin-2-ylamine, unii-d5qe8xw52u, 5-bromo-pyridin-2-ylamine, 2-amino-5-bromo pyridine, d5qe8xw52u
PubChem CID 70622
IUPAC Name 5-bromopyridin-2-amine
SMILES C1=CC(=NC=C1Br)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC153740050
SDS View Documents
Thermo Scientific Chemicals
153740050
5 g
Each for $59.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1072-97-5
173.01
WGOLHUGPTDEKCF-UHFFFAOYSA-N
70622
C1=CC(=NC=C1Br)N
C5H5BrN2
MFCD00006323
2-amino-5-bromopyridine, 2-pyridinamine, 5-bromo, 5-bromo-2-pyridylamine, 5-bromo-2-aminopyridine, 5-bromo-2-pyridinamine, 5-bromopyridin-2-ylamine, unii-d5qe8xw52u, 5-bromo-pyridin-2-ylamine, 2-amino-5-bromo pyridine, d5qe8xw52u
5-bromopyridin-2-amine

Specifications

1072-97-5
100.0
White to Beige
96% min. (GC)
C5H5BrN2
MFCD00006323
22, 431
WGOLHUGPTDEKCF-UHFFFAOYSA-N
5-bromopyridin-2-amine
70622
97%
2-Amino-5-bromopyridine, 97%
96.0
135.0°C to 138.0°C
Authentic
Glass bottle
Br(C5H3N)NH2
5 g
2-amino-5-bromopyridine, 2-pyridinamine, 5-bromo, 5-bromo-2-pyridylamine, 5-bromo-2-aminopyridine, 5-bromo-2-pyridinamine, 5-bromopyridin-2-ylamine, unii-d5qe8xw52u, 5-bromo-pyridin-2-ylamine, 2-amino-5-bromo pyridine, d5qe8xw52u
C1=CC(=NC=C1Br)N
173.01
173.01
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-019-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only