missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyridine, 97%

Catalog No. AC153740250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153740250 25 g
AC153740050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153740250 Supplier Thermo Scientific Chemicals Supplier No. 153740250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1072-97-5 | C5H5BrN2 | 173.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-97-5
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD00006323
InChI Key WGOLHUGPTDEKCF-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyridine, 2-pyridinamine, 5-bromo, 5-bromo-2-pyridylamine, 5-bromo-2-aminopyridine, 5-bromo-2-pyridinamine, 5-bromopyridin-2-ylamine, unii-d5qe8xw52u, 5-bromo-pyridin-2-ylamine, 2-amino-5-bromo pyridine, d5qe8xw52u
PubChem CID 70622
IUPAC Name 5-bromopyridin-2-amine
SMILES C1=CC(=NC=C1Br)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 135.0°C to 138.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br(C5H3N)NH2
Quantity 25 g
Beilstein 22, 431
Formula Weight 173.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromopyridine, 97%
Show More Show Less
EINECSNumber 214-019-9
TSCA TSCA

RUO – Research Use Only