missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog No. AC432500050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432500050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432500050 Supplier Thermo Scientific Chemicals Supplier No. 432500050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 944401-56-3
Molecular Formula C6H4BrF3N2
Molecular Weight (g/mol) 241.01
MDL Number MFCD09864896
InChI Key AEWYLVDLWQPJGW-UHFFFAOYSA-N
Synonym 5-bromo-4-trifluoromethyl pyridin-2-amine, 2-amino-5-bromo-4-trifluoromethyl pyridine, 5-bromo-4-trifluoromethyl-2-pyridylamine, 2-amino-5-bromo-4-trifluoromethylpyridine, acmc-209rru, 2-amino-4-trifluoromethyl-5-bromopyridine, 5-bromo-4-trifluoromethyl-2-pyridinamine, 5-bromo-4-trifluoromethyl-2-pyridinylamine, 5-bromanyl-4-trifluoromethyl pyridin-2-amine
PubChem CID 28875348
IUPAC Name 5-bromo-4-(trifluoromethyl)pyridin-2-amine
SMILES NC1=NC=C(Br)C(=C1)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 71°C to 74°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 241.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-4-(trifluoromethyl)pyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remov

RUO – Research Use Only