missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-methylpyridine 98.0+%, TCI America™

Catalog No. p-7117621
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18971G 1 g
A18975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18971G Supplier TCI America Supplier No. A18971G
Only null left

Chemical Identifiers

CAS 98198-48-2
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
InChI Key JDNCMHOKWINDKI-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-4-methylpyridine, 2-amino-4-methyl-5-bromopyridine, 2-amino-5-bromo-4-picoline, 5-bromo-4-methyl-pyridin-2-ylamine, 5-bromo-4-methyl-2-pyridinamine, 5-bromo-4-methyl-2-pyridinylamine, 2-pyridinamine, 5-bromo-4-methyl, 5-bromo-4-methyl-2-pyridylamine, 2-amino-5-bromo-4-picoline 2-amino-5-bromo-4-methylpyridine, pubchem2241
PubChem CID 817695
IUPAC Name 5-bromo-4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1Br)N

Specifications

Melting Point 151°C
Quantity 1 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-4-methylpyridine