missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-bromo-3-nitropyridine 97.0+%, TCI America™
$80.16 - $240.46
Chemical Identifiers
| CAS | 6945-68-2 |
|---|---|
| Molecular Formula | C5H4BrN3O2 |
| Molecular Weight (g/mol) | 218.01 |
| MDL Number | MFCD00047441 |
| InChI Key | QOOCOFOGYRQPPN-UHFFFAOYSA-N |
| Synonym | 2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b |
| PubChem CID | 138878 |
| IUPAC Name | 5-bromo-3-nitropyridin-2-amine |
| SMILES | NC1=NC=C(Br)C=C1[N+]([O-])=O |
Chemical Identifiers
| 6945-68-2 | |
| 218.01 | |
| QOOCOFOGYRQPPN-UHFFFAOYSA-N | |
| 138878 | |
| NC1=NC=C(Br)C=C1[N+]([O-])=O |
| C5H4BrN3O2 | |
| MFCD00047441 | |
| 2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b | |
| 5-bromo-3-nitropyridin-2-amine |
Specifications
| 6945-68-2 | |
| Yellow | |
| MFCD00047441 | |
| 2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b | |
| NC1=NC=C(Br)C=C1[N+]([O-])=O | |
| 218.01 | |
| 218.01 | |
| Crystalline Powder |
| 209°C | |
| C5H4BrN3O2 | |
| 5 g | |
| QOOCOFOGYRQPPN-UHFFFAOYSA-N | |
| 5-bromo-3-nitropyridin-2-amine | |
| 138878 | |
| ≥97.0% (GC) | |
| 2-Amino-5-bromo-3-nitropyridine |
Safety and Handling
TSCA : No