missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-nitropyridine 97.0+%, TCI America™

Catalog No. A15565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15565G 5 g
A155625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15565G Supplier TCI America Supplier No. A15565G
Only null left

Chemical Identifiers

CAS 6945-68-2
Molecular Formula C5H4BrN3O2
Molecular Weight (g/mol) 218.01
MDL Number MFCD00047441
InChI Key QOOCOFOGYRQPPN-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
PubChem CID 138878
IUPAC Name 5-bromo-3-nitropyridin-2-amine
SMILES NC1=NC=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 209°C
Color Yellow
Quantity 5 g
Formula Weight 218.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-3-nitropyridine