missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-nitropyridine, 97%

CAS: 6945-68-2 | C5H4BrN3O2 | 218.01 g/mol

$70.97 - $236.55

Chemical Identifiers

CAS 6945-68-2
Molecular Formula C5H4BrN3O2
Molecular Weight (g/mol) 218.01
MDL Number MFCD00047441
InChI Key QOOCOFOGYRQPPN-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
PubChem CID 138878
IUPAC Name 5-bromo-3-nitropyridin-2-amine
SMILES NC1=NC=C(Br)C=C1[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1498506
View Documents
Thermo Scientific Chemicals
A1498506
5 g
Each for $70.97
Only null left
 
AAA1498514
View Documents
Thermo Scientific Chemicals
A1498514
25 g
Each for $236.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6945-68-2
218.01
QOOCOFOGYRQPPN-UHFFFAOYSA-N
138878
NC1=NC=C(Br)C=C1[N+]([O-])=O
C5H4BrN3O2
MFCD00047441
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
5-bromo-3-nitropyridin-2-amine

Specifications

6945-68-2
C5H4BrN3O2
5 g
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
NC1=NC=C(Br)C=C1[N+]([O-])=O
218.01
218.01
2-Amino-5-bromo-3-nitropyridine
206°C to 210°C
MFCD00047441
383851
QOOCOFOGYRQPPN-UHFFFAOYSA-N
5-bromo-3-nitropyridin-2-amine
138878
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only