missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-methylpyridine 98.0+%, TCI America™

Catalog No. A21541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21541G 1 g
A21545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21541G Supplier TCI America Supplier No. A21541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3430-21-5 | C6H7BrN2 | 187.04 g/mol

Chemical Identifiers

CAS 3430-21-5
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD00068232
InChI Key KBLGGRWUEVCNPY-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
PubChem CID 137934
IUPAC Name 5-bromo-3-methylpyridin-2-amine
SMILES CC1=CC(=CN=C1N)Br

Specifications

Melting Point 94°C
Quantity 1 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-3-methylpyridine