missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-methylpyridine, 97%

Catalog No. AAA1531806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1531806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1531806 Supplier Thermo Scientific Chemicals Supplier No. A1531806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3430-21-5 | C6H8BrN2 | 188.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 3430-21-5
MDL Number MFCD00068232
Synonym 2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
PubChem CID 137934

Specifications

Melting Point 90°C to 94°C
Odor Unpleasant
Molecular Formula C6H7BrN2
Quantity 5 g
Beilstein 386426
Sensitivity Hygroscopic
InChI Key KBLGGRWUEVCNPY-UHFFFAOYSA-N
SMILES CC1=CC(=CN=C1N)Br
IUPAC Name 5-bromo-3-methylpyridin-2-amine
Molecular Weight (g/mol) 187.04
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3-methylpyridine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only