missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3,4-dimethylpyridine, 97%

Catalog No. p-7047126
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1767203 1 g
AAL1767206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1767203 Supplier Thermo Scientific Chemicals Supplier No. L1767203
Only null left

CAS: 374537-97-0 | C7H9BrN2 | 201.07 g/mol

It is an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 374537-97-0
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.07
MDL Number MFCD02093947
InChI Key YAVKJNIMFGZBSY-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3,4-dimethylpyridine, 5-bromo-3,4-dimethyl-2-pyridinamine, acmc-20agep, 5-bromo-3,4-dimethyl-2-pyridylamine, 2-pyridinamine,5-bromo-3,4-dimethyl, 5-bromo-3,4-dimethyl-pyridin-2-ylamine
PubChem CID 7010307
IUPAC Name 5-bromo-3,4-dimethylpyridin-2-amine
SMILES CC1=C(C)C(N)=NC=C1Br

Specifications

Melting Point 133°C to 135°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 201.07
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3,4-dimethylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only