missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5,6-dimethylbenzothiazole 98.0+%, TCI America™
$436.19 - $1735.43
Chemical Identifiers
| CAS | 29927-08-0 |
|---|---|
| Molecular Formula | C9H10N2S |
| Molecular Weight (g/mol) | 178.25 |
| MDL Number | MFCD00005790 |
| InChI Key | IODWHFFPQHUDAG-UHFFFAOYSA-N |
| PubChem CID | 34757 |
| IUPAC Name | 5,6-dimethyl-1,3-benzothiazol-2-amine |
| SMILES | CC1=C(C)C=C2N=C(N)SC2=C1 |
Chemical Identifiers
| 29927-08-0 | |
| 178.25 | |
| IODWHFFPQHUDAG-UHFFFAOYSA-N | |
| 5,6-dimethyl-1,3-benzothiazol-2-amine |
| C9H10N2S | |
| MFCD00005790 | |
| 34757 | |
| CC1=C(C)C=C2N=C(N)SC2=C1 |
Specifications
| 29927-08-0 | |
| C9H10N2S | |
| 5 g | |
| IODWHFFPQHUDAG-UHFFFAOYSA-N | |
| 5,6-dimethyl-1,3-benzothiazol-2-amine | |
| 34757 | |
| ≥98.0% (HPLC,T) | |
| 2-Amino-5,6-dimethylbenzothiazole |
| 190°C | |
| MFCD00005790 | |
| 2-amino-5,6-dimethylbenzothiazole, 5,6-dimethylbenzo d thiazol-2-amine, 2-benzothiazolamine, 5,6-dimethyl, 5,6-dimethyl-2-aminobenzothiazole, unii-6ybu8z59ys, 5,6-dimethylbenzothiazol-2-ylamine, 6ybu8z59ys, 5,6-dimethyl-2-benzothiazolamine, 2-benzothiazolamine,5,6-dimethyl, 2-amino-5,6-dimethyl-1,3-benzothiazol | |
| CC1=C(C)C=C2N=C(N)SC2=C1 | |
| 178.25 | |
| 178.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No