missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5,6-dimethylbenzothiazole 98.0+%, TCI America™

Catalog No. A14615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14615G 5 g
A146125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14615G Supplier TCI America Supplier No. A14615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29927-08-0
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.25
MDL Number MFCD00005790
InChI Key IODWHFFPQHUDAG-UHFFFAOYSA-N
PubChem CID 34757
IUPAC Name 5,6-dimethyl-1,3-benzothiazol-2-amine
SMILES CC1=C(C)C=C2N=C(N)SC2=C1

Specifications

Melting Point 190°C
Quantity 5 g
Synonym 2-amino-5,6-dimethylbenzothiazole, 5,6-dimethylbenzo d thiazol-2-amine, 2-benzothiazolamine, 5,6-dimethyl, 5,6-dimethyl-2-aminobenzothiazole, unii-6ybu8z59ys, 5,6-dimethylbenzothiazol-2-ylamine, 6ybu8z59ys, 5,6-dimethyl-2-benzothiazolamine, 2-benzothiazolamine,5,6-dimethyl, 2-amino-5,6-dimethyl-1,3-benzothiazol
Formula Weight 178.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5,6-dimethylbenzothiazole