missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

Catalog No. AAH3205906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3205906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3205906 Supplier Thermo Scientific Chemicals Supplier No. H3205906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 70291-62-2 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70291-62-2
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00121495
InChI Key RILAXFOADRDGPQ-UHFFFAOYSA-N
PubChem CID 258182
IUPAC Name 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
SMILES NC1=C(C#N)C2=C(CCC2)S1

Specifications

Melting Point 153°C to 154°C
Quantity 5 g
UN Number UN3439
Synonym 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4,5-trimethylene-3-thiophenecarbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-5,6-dihydro, 2-amino-3-cyano-5,6-dihydro-4h-cyclopenta b thiophene, 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile,2-amino-5,6-dihydro, maybridge1_003206, 4,5-trimethylene-2-aminothiophene-3-carbonitrile, 2-amino-5,6-dihydro-4h-cyclopenta b thioph ene-3-carbonitrile
Formula Weight 164.23
Chemical Name or Material 2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only