missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 98.0+%, TCI America™

Catalog No. p-7118213
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30871G 1 g
A30875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30871G Supplier TCI America Supplier No. A30871G

Chemical Identifiers

CAS 402960-38-7
Molecular Formula C10H16BN3O2
Molecular Weight (g/mol) 221.067
MDL Number MFCD06795685
InChI Key BPQVMIDUTRJYSC-UHFFFAOYSA-N
Synonym 2-aminopyrimidine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-aminopyrimidine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-amino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrimidin-2-amine, 2-aminopyrimidin-5-yl boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinylamine, 2-2-aminopyrimidin-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 17750212
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)N

Specifications

Melting Point 212°C
Quantity 1 g
Formula Weight 221.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine