missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one 98.0+%, TCI America™
$70.84 - $359.76
Chemical Identifiers
| CAS | 105355-26-8 |
|---|---|
| Molecular Formula | C19H21N3O2S |
| Molecular Weight (g/mol) | 355.456 |
| MDL Number | MFCD07368627 |
| InChI Key | ABGJULHDDNEULW-UHFFFAOYSA-N |
| Synonym | 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2-imino-4-thiazolidinone |
| PubChem CID | 11416954 |
| IUPAC Name | 2-amino-5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazol-4-one |
| SMILES | CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)N=C(S3)N |
Chemical Identifiers
| 105355-26-8 | |
| 355.456 | |
| ABGJULHDDNEULW-UHFFFAOYSA-N | |
| 11416954 | |
| CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)N=C(S3)N |
| C19H21N3O2S | |
| MFCD07368627 | |
| 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2-imino-4-thiazolidinone | |
| 2-amino-5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazol-4-one |
Specifications
| 105355-26-8 | |
| C19H21N3O2S | |
| 5 g | |
| ABGJULHDDNEULW-UHFFFAOYSA-N | |
| 2-amino-5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazol-4-one | |
| 11416954 | |
| ≥98.0% (HPLC,T) | |
| 2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one |
| 196°C | |
| MFCD07368627 | |
| 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2-imino-4-thiazolidinone | |
| CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)N=C(S3)N | |
| 355.456 | |
| 355.46 | |
| Crystalline Powder |
Safety and Handling
TSCA : No