missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 95%

Catalog No. p-7036124
Change view
Click to view available options
Quantity:
1 g
250 mg
Catalog No. Quantity
AAH54372MD 250 mg
AAH5437203 1 g
2 options

Catalog No. AAH54372MD

Supplier: Thermo Scientific Chemicals H54372MD

Only null left

CAS: 944401-57-4 | C12H16BF3N2O2 | 288.077 g/mol

2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 944401-57-4
Molecular Formula C12H16BF3N2O2
Molecular Weight (g/mol) 288.077
MDL Number MFCD12923420
InChI Key AHNBKJSRXQDYEO-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4-trifluoromethyl pyridin-2-amine, 2-amino-4-trifluoromethyl pyridine-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-4-trifluoromethyl pyridin-2-amine, 6-amino-4-trifluoromethyl pyridin-3-yl boronic acid pinacol ester, 4-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 4-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridm-2-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-4-trifluoromethyl pyridin-2-amine
PubChem CID 57416499
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2C(F)(F)F)N

Specifications

Quantity 250 mg
Beilstein 15519102
Solubility Information Slightly soluble in water.
Formula Weight 288.07
Percent Purity 95%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only