missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-tert-butylthiazole, 98%

CAS: 74370-93-7 | C7H12N2S | 156.247 g/mol

Supplier:  Thermo Scientific Chemicals A1411506

 View more versions of this product

Catalog No. AAA1411506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-4-tert-butylthiazole
98°C to 103°C
MFCD00051644
114270
CUWZBHVYLVGOAB-UHFFFAOYSA-N
4-tert-butyl-1,3-thiazol-2-amine
2734202
98%
74370-93-7
C7H12N2S
5 g
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
CC(C)(C)C1=CSC(=N1)N
156.247
156.26

Chemical Identifiers

74370-93-7
156.247
CUWZBHVYLVGOAB-UHFFFAOYSA-N
2734202
CC(C)(C)C1=CSC(=N1)N
C7H12N2S
MFCD00051644
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
4-tert-butyl-1,3-thiazol-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c

H302-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only